Skip to Content
Merck
All Photos(1)

Key Documents

465313

Sigma-Aldrich

1-Methylindole-3-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

197-200 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

Cn1cc(C(O)=O)c2ccccc12

InChI

1S/C10H9NO2/c1-11-6-8(10(12)13)7-4-2-3-5-9(7)11/h2-6H,1H3,(H,12,13)

InChI key

HVRCLXXJIQTXHC-UHFFFAOYSA-N

General description

1-Methylindole-3-carboxylic acid is an indole derivative that can be prepared by the oxidation of 1-methylindole-3-aldehyde with alkaline potassium permanganate.[1][2]

Application

  • Reactant for preparation of bisindolyl pyrimidinones analogs of PKC inhibitor LY333531
  • Reactant for preparation of (heteroaryl)(carboxamido)arylpyrrole derivatives as Cdc7 kinase inhibitors, antitumor and antiproliferative agents
  • Reactant for preparation of (pyrrolidinylmethoxy)cyclohexanecarboxylic acids as antigen-4 (VLA-4) antagonists
  • Reactant for preparation of EphB3 receptor tyrosine kinase inhibitors
  • Reactant for preparation of pyrazolodiazepine derivatives as human P2X7 receptor antagonists
  • Reactant for preparation of potent nonpeptidic urotensin II receptor agonists
  • Reactant for preparation of pyrrolizidine esters, amides, and ureas as 5-HT4 receptor ligands

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction of Some Indole Ketones with Iodine and Pyridine.
Hart G and Potts KT.
The Journal of Organic Chemistry, 27(8), 2940-2942 (1962)
Desulfurization of Thiiranes with Iodine.
Helmkamp GK and Pettitt DJ.
The Journal of Organic Chemistry, 27(80, 2942-2943 (1962)
Mitra Shokrollahi et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 31(10), e13598-e13598 (2019-04-24)
Activating luminal 5-HT4 receptors results in the release of 5-HT from enterochromaffin cells into the lamina propria to modulate colonic motility. Our aim was to evaluate characteristics of colonic motor patterns involved in the prokinetic effects of intraluminal prucalopride in
Yuan Guo et al.
Neuropsychobiology, 80(1), 52-63 (2020-07-15)
The 5-hydroxytryptamine (5-HT) neurotransmitter system and lateral habenula (LHb) are involved in the regulation of depression, while the mechanisms remain to be clarified. The effects and possible mecha-nism underlying activation or blockade of 5-HT4 receptors (5-HT4Rs) in the LHb in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service