Skip to Content
Merck
All Photos(1)

Key Documents

465127

Sigma-Aldrich

N-(tert-Butoxycarbonyl)glycine N′-methoxy-N′-methylamide

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$279.00

$279.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
$279.00

About This Item

Linear Formula:
(CH3)3COCONHCH2CON(OCH3)CH3
CAS Number:
Molecular Weight:
218.25
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$279.00


Estimated to ship on30 May 2025


Request a Bulk Order

Assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

mp

100-104 °C (lit.)

application(s)

peptide synthesis

SMILES string

CON(C)C(=O)CNC(=O)OC(C)(C)C

InChI

1S/C9H18N2O4/c1-9(2,3)15-8(13)10-6-7(12)11(4)14-5/h6H2,1-5H3,(H,10,13)

InChI key

XJVZHKXGDQXSNT-UHFFFAOYSA-N

Application

α-Amino Weinreb amide.[1][2][3][4][5][6][7][8]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucet, D. et al.
Tetrahedron Asymmetry, 7, 985-985 (1996)
Kosynkina, L. et al.
Tetrahedron Letters, 35, 5173-5173 (1994)
Kim, B.M. et al.
Tetrahedron Letters, 35, 5153-5153 (1994)
Overhand, M. Hecht, S.M
The Journal of Organic Chemistry, 59, 4721-4721 (1994)
S. Nahm et al.
Tetrahedron Letters, 22, 3815-3815 (1981)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service