Skip to Content
Merck
All Photos(1)

Key Documents

464678

Sigma-Aldrich

Palladium(II) iodide

97%

Synonym(s):

Palladium diiodide, Palladium iodide, Palladous iodide

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$291.00

$291.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
$291.00

About This Item

Empirical Formula (Hill Notation):
I2Pd
CAS Number:
Molecular Weight:
360.23
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

$291.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: palladium

mp

350 °C (dec.) (lit.)

density

6.003 g/mL at 25 °C (lit.)

SMILES string

I[Pd]I

InChI

1S/2HI.Pd/h2*1H;/q;;+2/p-2

InChI key

HNNUTDROYPGBMR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Method: Formation of a red-violet dye with chrom azurol S.

Measuring ranges may vary when used in combination with other photometers or methods than specified above.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service