Skip to Content
Merck
All Photos(2)

Key Documents

464120

Sigma-Aldrich

3,4-Dihydroxy-5-methoxybenzaldehyde

96%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$125.00
5 G
$395.00

$125.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$125.00
5 G
$395.00

About This Item

Linear Formula:
CH3OC6H2(OH)2CHO
CAS Number:
Molecular Weight:
168.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$125.00


Available to ship on22 April 2025Details


Request a Bulk Order

Assay

96%

mp

131-134 °C (lit.)

functional group

aldehyde

SMILES string

COc1cc(C=O)cc(O)c1O

InChI

1S/C8H8O4/c1-12-7-3-5(4-9)2-6(10)8(7)11/h2-4,10-11H,1H3

InChI key

RRKMWVISRMWBAL-UHFFFAOYSA-N

General description

3,4-Dihydroxy-5-methoxybenzaldehyde can be obtained by reactting 5-iodovaniliin with sodium hydroxide and copper sulfate solution.[1]

Application

3,4-Dihydroxy-5-methoxybenzaldehyde may be used for the preparation of 3,4-dihydroxy-6-methoxy-β-nitrostyrene[2] and 5-hydroxyconiferyl alcohol.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and synthesis of (?)-Wuweizisu C.
Schneiders GE and Stevenson R.
The Journal of Organic Chemistry, 46(41), 2969-2971 (1981)
Thomas Goujon et al.
Plant molecular biology, 51(6), 973-989 (2003-06-05)
A promoter-trap screen allowed us to identify an Arabidopsis line expressing GUS in the root vascular tissues. T-DNA border sequencing showed that the line was mutated in the caffeic acid O-methyltransferase 1 gene (AtOMT1) and therefore deficient in OMT1 activity.
MESCALINE ANALOGS. III. 2, 4, 6-TRIALKYL-AND 3, 4-DIHYDROXY-5-METHOXY-?-PHENETHYLAMINES.
Benington F, et al.
The Journal of Organic Chemistry, 20(9), 1292-1296 (1955)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service