Skip to Content
Merck
All Photos(1)

Key Documents

463027

Sigma-Aldrich

Germane

electronic grade, ≥99.997%

Synonym(s):

Germanium tetrahydride, Germanium(4+) hydride, Monogermane, Tetrahydrogermane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GeH4
CAS Number:
Molecular Weight:
76.67
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

electronic grade

vapor density

1.53 (−142 °C, vs air)

Assay

≥99.997%

form

gas

impurities

<0.5 ppm Oxygen (O2)
<1 ppm Carbon monoxide + Trigermane (CO+Ge3H8)
<1 ppm THC
<1 ppm Water (H2O)
<2 ppm Carbon dioxide (CO2)
<2 ppm Nitrogen (N2)
<20 ppm Digermane (Ge2H6)
<5 ppm Chlorogermanes
<5 ppm Germoxanes
<50 ppm Hydrogen (H2)

bp

−88.4 °C (lit.)

mp

−165 °C (lit.)

transition temp

critical temperature 35 °C

SMILES string

[GeH4]

InChI

1S/GeH4/h1H4

InChI key

QUZPNFFHZPRKJD-UHFFFAOYSA-N

Features and Benefits

Germane gas is used in the deposition of epitaxial and amorphous SiGe alloy layers used in the production of high performance devices including photovoltaic cells and integrated circuits (IC). Precursor to germanium-containing thin films by chemical vapor deposition (CVD) and molecular beam epitaxy (MBE). A CVD precursor for germanium carbide semiconducting films.

Recommended products

Stainless steel regulators Z527416 or Z527424 are recommended.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanjie Yang et al.
International journal of nanomedicine, 14, 6297-6311 (2019-09-10)
Mn-doped ZnS quantum dots (QDs) with special luminescent properties have been widely researched and applied in various fields. Thus, their release toxicity and security cannot be ignored. In the present study, the toxicity and non-targeted metabolomics of Mn-doped ZnS QDs
Woranud Pakdeedachakiat et al.
Bioresource technology, 291, 121873-121873 (2019-08-05)
Cholinium-glycinate ([Ch][Gly]) and cholinium-alanate ([Ch][Ala]) were investigated on the pretreatment of mulberry stem (MS). It resulted in an increase of glucose from 14% to more than 74% compared to the untreated sample. Pretreatment by reused [Ch][Gly] showed good performance for
Yun Fang et al.
Applied and environmental microbiology, 85(16) (2019-06-09)
Bacterial anaerobic respiration using an extracellular electron acceptor plays a predominant role in global biogeochemical cycles. However, the mechanisms of bacterial adaptation to the toxic organic pollutant as the extracellular electron acceptor during anaerobic respiration are not clear, which limits
Silvia C Richner et al.
Health and quality of life outcomes, 15(1), 56-56 (2017-03-28)
The regret intensity scale (RIS) and the regret coping scale for healthcare professionals (RCS-HCP) working in hospitals assess the experience of care-related regrets and how healthcare professional deal with these negative events. The aim of this study was to validate
Ran Zhang et al.
Nature communications, 10(1), 3760-3760 (2019-08-23)
Adeno-associated virus (AAV) receptor (AAVR) is an essential receptor for the entry of multiple AAV serotypes with divergent rules; however, the mechanism remains unclear. Here, we determine the structures of the AAV1-AAVR and AAV5-AAVR complexes, revealing the molecular details by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service