Skip to Content
Merck
All Photos(1)

Key Documents

462934

Sigma-Aldrich

Ethyl phenoxyacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CO2C2H5
CAS Number:
Molecular Weight:
180.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Assay

98%

refractive index

n20/D 1.505 (lit.)

bp

136 °C/19 mmHg (lit.)

density

1.1 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)COc1ccccc1

InChI

1S/C10H12O3/c1-2-12-10(11)8-13-9-6-4-3-5-7-9/h3-7H,2,8H2,1H3

InChI key

MGZFVSUXQXCEHM-UHFFFAOYSA-N

General description

Ethyl phenoxyacetate is an ester. It undergoes cyanide ion catalyzed aminolysis with ammonia to afford the corresponding amide.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Ethyl Phenoxyacetate Derivatives under Conditions of Phase-Transfer Catalysis and Ultrasonic Irradiation.
Kukovinets AG, et al.
Russ. J. Org. Chem., 34, 1331-1333 (1998)
Cyanide as an efficient and mild catalyst in the aminolysis of esters
Hogberg T, et al.
The Journal of Organic Chemistry, 52, 2033-2036 (1987)
Yongqiang Wu et al.
Luminescence : the journal of biological and chemical luminescence, 33(5), 855-862 (2018-04-18)
Six novel 8-hydroxyquinoline derivatives were synthesized using 2-methyl-8-hydroxyquinoline and para-substituted phenol as the main starting materials, and were characterized by 1 H nuclear magnetic resonance (NMR), mass spectrometry (MS), ultraviolet (UV) light analysis and infra-red (IR) light analysis. Their complexes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service