Skip to Content
Merck
All Photos(1)

Documents

462896

Sigma-Aldrich

Poly(styrene-co-methyl methacrylate)

average Mw 100,000-150,000, pellets, styrene ~40 mol %

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

pellets

melt index

1.0 g/10 min (200°C, 5.0kg)

mol wt

average Mw 100,000-150,000

composition

styrene, ~40 mol %

transition temp

Tg (DSC) 101 °C (onset)

SMILES string

COC(=O)C(C)=C.C=Cc1ccccc1

InChI

1S/C8H8.C5H8O2/c1-2-8-6-4-3-5-7-8;1-4(2)5(6)7-3/h2-7H,1H2;1H2,2-3H3

InChI key

ADFPJHOAARPYLP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M E Forsythe et al.
Journal of orthopaedic surgery (Hong Kong), 14(1), 38-42 (2006-04-07)
To compare the pharmacokinetic profile of tobramycin in blood, urine, and at the operative site following the use of Simplex-tobramycin bone cement in primary total hip replacement between patients with and without renal dysfunction. Six patients with renal dysfunction underwent
N S Dar-Odeh et al.
Journal of oral rehabilitation, 24(10), 755-760 (1997-12-31)
The aim of this study was to compare some of the physical properties of a heat-cured, a self-cured and a visible light-cured acrylic resin, and to evaluate the suitability of visible light-cured resin as a repair material for dentures made
G N Thomopoulos et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 31(7), 871-878 (1983-07-01)
The high iron diamine (HID) method for detection of sulfated complex carbohydrate has been applied directly on thin sections of variably fixed tissues embedded in epoxy and nonepoxy resins. Results with postembedment HID staining in mouse intestinal epithelium are compared
MN: Surgical LPN files product liability suit: cement Mfgr.'s failure to warn of health hazard.
Nursing law's Regan report, 42(8), 3-3 (2004-08-11)
Novel fabrication of 2D and 3D inverted opals and their application.
Jae Ah Lee et al.
Small (Weinheim an der Bergstrasse, Germany), 7(18), 2581-2586 (2011-08-03)

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service