Skip to Content
Merck
All Photos(1)

Key Documents

462314

Sigma-Aldrich

Iminodi(methylphosphonic acid)

97%

Synonym(s):

Aminobis(methylenephosphonic acid), Bis(phosphonomethyl)amine, Iminobis(methylene)bis(phosphonic acid), Iminobis[methylenephosphonic acid], Iminodimethylenediphosphonic acid, Iminodimethylenephosphonic acid, P,P′-[Iminobis(methylene)]bis[phosphonic acid]

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$132.00

$132.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$132.00

About This Item

Linear Formula:
HN[CH2PO(OH)2]2
CAS Number:
Molecular Weight:
205.04
Beilstein:
1812882
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$132.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

220 °C (dec.) (lit.)

solubility

water: soluble

functional group

amine

SMILES string

OP(O)(=O)CNCP(O)(O)=O

InChI

1S/C2H9NO6P2/c4-10(5,6)1-3-2-11(7,8)9/h3H,1-2H2,(H2,4,5,6)(H2,7,8,9)

InChI key

ISQSUCKLLKRTBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iminodi(methylphosphonic acid) is an iminopolyphosphonic acid derivative. It is widely used as complex forming reagent for alkylphosphonic groups. Thermal analysis has confirmed the anhydrous state of iminodi(methylphosphonic acid).[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iminodi (methylphosphonic) acid.
Afonin, E. G.
Russ. J. Gen. Chem., 73(10), 1503-1505 (2003)
Scott A Lujan et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(30), 12282-12287 (2007-07-17)
Conjugative transfer of plasmid DNA via close cell-cell junctions is the main route by which antibiotic resistance genes spread between bacterial strains. Relaxases are essential for conjugative transfer and act by cleaving DNA strands and forming covalent phosphotyrosine linkages. Based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service