Skip to Content
Merck
All Photos(2)

Key Documents

462233

Sigma-Aldrich

D-Galactal

95%

Synonym(s):

1,5-Anhydro-2-deoxy-D-lyxo-hex-1-enitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O4
CAS Number:
Molecular Weight:
146.14
Beilstein:
81690
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

optical activity

[α]22/D −21.5°, c = 1.2 in methanol

mp

99-103 °C (lit.)

storage temp.

2-8°C

SMILES string

OC[C@H]1OC=C[C@@H](O)[C@H]1O

InChI

1S/C6H10O4/c7-3-5-6(9)4(8)1-2-10-5/h1-2,4-9H,3H2/t4-,5-,6-/m1/s1

InChI key

YVECGMZCTULTIS-HSUXUTPPSA-N

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aditi Agarwal et al.
The Journal of organic chemistry, 69(18), 6137-6140 (2004-09-18)
Perchloric acid supported on silica gel acts as an excellent reagent system in converting glucals into 2,3-unsaturated-O-glucosides in good to excellent yields in short reaction time with good alpha selectivity. Primary, secondary, and allylic alcohols, phenols, and thiols react with
Chi-Li Chen et al.
Organic letters, 11(2), 409-412 (2009-01-09)
With focus on the steric effects present in the transition states for the [3,3]-sigmatropic rearrangement, the substrate 5 has been designed to improve the overall stereoselectivity of the Ireland-Claisen rearrangement. Experimentally, it has been found that (1) only Z-6 rearranges
Feiqing Ding et al.
Organic letters, 13(4), 652-655 (2011-01-20)
A highly efficient synthesis of L-ristosamine and L-epi-daunosamine glycosides via BF(3)·OEt(2) promoted tandem hydroamination/glycosylation of 3,4-di-O-acetyl-6-deoxy-L-glucal and L-galactal has been developed. The new method proceeds in a completely stereocontrolled manner within a short reaction time. Preparation of a library of
Jürgen Geiger et al.
The Journal of organic chemistry, 72(12), 4367-4377 (2007-05-17)
A 3,4-O-unprotected galactal derivative having bulky 6-O-TIPS protection (compound 2) could be regioselectively 3-O-glycosylated with O-(galactopyranosyl) trichloroacetimidates; depending on the protecting group pattern stereoselectively alpha- and beta-linked disaccharides were obtained. With O-(2-azido-2-deoxyglucopyransyl) trichloroacetimidate as donor (compound 10A), glycosylation of 2
Y X Lian et al.
Zhongguo Zhong xi yi jie he za zhi Zhongguo Zhongxiyi jiehe zazhi = Chinese journal of integrated traditional and Western medicine, 21(9), 649-651 (2003-02-11)
To observe the effect of Shuangcao Tuihuang Granule-1 (SCTH-1) in treating severe jaundice of acute icterohepatitis and to study its mechanism. Thirty-four patients with severe jaundice of acute icterohepatitis were treated with SCTH-1, their therapeutic effects were analyzed. In the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service