Skip to Content
Merck
All Photos(1)

Key Documents

462152

Sigma-Aldrich

Methyl 4-iodobutyrate

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$179.00

$179.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$179.00

About This Item

Linear Formula:
I(CH2)3CO2CH3
CAS Number:
Molecular Weight:
228.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$179.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

contains

copper as stabilizer

refractive index

n20/D 1.505 (lit.)

bp

80-83 °C/11 mmHg (lit.)

density

1.689 g/mL at 25 °C (lit.)

functional group

ester
iodo

storage temp.

2-8°C

SMILES string

COC(=O)CCCI

InChI

1S/C5H9IO2/c1-8-5(7)3-2-4-6/h2-4H2,1H3

InChI key

NBCIIVXSBPDKOM-UHFFFAOYSA-N

General description

Methyl 4-iodobutyrate can be synthesized from methyl-4-chlorobutyrate and sodium iodide.[1]

Application

Methyl 4-iodobutyrate may be used as reagent in the synthesis of 4-[formyl-5-(methoxymethyl)-1H-pyrrol-1-yl] butanoic acid.[2] It has been used in the synthesis of a range of stable S-adenosylmethionine (SAM) mimetics. Their ability to promote the binding of the E. coli methionine repressor (MetJ) to its operator DNA has been investigated.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young-Won Chin et al.
Bioorganic & medicinal chemistry letters, 13(1), 79-81 (2002-12-07)
As a part of our search for hepatoprotective compounds from Lycium chinense fruits, three new pyrrole derivatives (1-3) were isolated. These compounds and a related synthetic methylated compound (4) were evaluated for their biological activity and structure-activity relationship, and compounds
Catherine Joce et al.
Organic & biomolecular chemistry, 7(4), 635-638 (2009-02-06)
The efficient synthesis of a range of stable SAM mimetics, and their ability to promote the binding of the E. coli methionine repressor (MetJ) to its operator DNA, is described.
IgE-mediated immediate-type hypersensitivity to the pyrazolone drug propyphenazone.
Himly M, et al.
The Journal of Allergy and Clinical Immunology, 111(4), 882-888 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service