Skip to Content
Merck
All Photos(1)

Key Documents

462047

Sigma-Aldrich

1-(2,6-Dimethylphenoxy)-2-propanamine hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3OCH2CH(CH3)NH2·HCl
CAS Number:
Molecular Weight:
215.72
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

200-203 °C (lit.)

SMILES string

Cl.CC(N)COc1c(C)cccc1C

InChI

1S/C11H17NO.ClH/c1-8-5-4-6-9(2)11(8)13-7-10(3)12;/h4-6,10H,7,12H2,1-3H3;1H

InChI key

NFEIBWMZVIVJLQ-UHFFFAOYSA-N

General description

1-(2,6-Dimethylphenoxy)-2-propanamine hydrochloride (Mexiletine) is an anti-arrhythmic agent having local anaesthetic activity.[1] Mexiletine belongs to the triclinc crystal system, with the asymmetric unit containing two independent molecules.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sivy J, et al.
Acta Crystallographica Section C, Structural Chemistry, 47(12), 2695-2695 (1991)
Erythromelagia: a rare and hard-to-treat condition: a 9-year-old boy responsive to intravenous lidocaine and oral mexilitene.
Francisca Elgueta et al.
Pain medicine (Malden, Mass.), 14(2), 311-312 (2013-02-02)
Lushan Yu et al.
The Journal of pharmacy and pharmacology, 64(6), 792-801 (2012-05-11)
This study examined the interaction of mexiletine enantiomers with human plasma, human serum albumin (HSA), and human α1-acid glycoprotein (hAGP), and characterized the binding modes of mexiletine enantiomers with hAGP in the molecular level. Enantiomer separation of mexiletine was performed
Tomoyuki Suzuki et al.
Cardiovascular research, 98(1), 136-144 (2013-01-24)
Na(+) channel blockers are often used to treat atrial fibrillation (AF), but may sometimes cause ventricular contractile dysfunction. However, amiodarone, a multi-channel blocker with Na(+) channel block, causes less contractile dysfunction. In this study, we tested the hypothesis that Na(+)
Yuanfeng Gao et al.
Circulation. Arrhythmia and electrophysiology, 6(3), 614-622 (2013-04-13)
Timothy syndrome (TS) is a rare long-QT syndrome caused by CACNA1C mutations G406R in exon 8A (TS1) and G402S/G406R in exon 8 (TS2). Management of TS is a challenge and prognosis is poor. This study aimed to explore the inheritance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service