Skip to Content
Merck
All Photos(1)

Key Documents

46195

Sigma-Aldrich

Farnesylacetone

technical, mixture of stereo isomers, ≥90% (GC)

Synonym(s):

6,10,14-Trimethyl-5,9,13-pentadecatrien-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H30O
CAS Number:
Molecular Weight:
262.43
Beilstein:
1781239
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (GC)

refractive index

n20/D 1.481

density

0.88 g/mL at 20 °C (lit.)

SMILES string

C\C(C)=C\CCC(C)=CCCC(C)=CCCC(C)=O

InChI

1S/C18H30O/c1-15(2)9-6-10-16(3)11-7-12-17(4)13-8-14-18(5)19/h9,11,13H,6-8,10,12,14H2,1-5H3

InChI key

LTUMRKDLVGQMJU-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geonseek Ryu et al.
Archives of pharmacal research, 26(10), 796-799 (2003-11-12)
Two known farnesylacetone derivatives (1 and 2) were isolated from the Korean brown alga Sargassum sagamianum off Jeju Island, Korea. Compounds 1 and 2 were identified as (5E,10Z)-6,10,14-trimethylpentadeca-5,10-dien-2,12-dione and (5E,9E,13E)-6,10,4-trimethyl-pentadeca-5,9,13-trien-2,12-dione, respectively, by comparison with the literature data. Compounds 1 and
Substrate specificity of undecaprenyl pyrophophate synthetase from Lactobacillus plantarum.
Baba T and Allen Jr CA.
Biochemistry, 17(26), 5598-5604 (1978)
Byong-Gon Park et al.
Bioorganic & medicinal chemistry letters, 18(24), 6324-6326 (2008-11-14)
Two farnesylacetones, 311 and 312, major active constituents of Sargassum siliquastrum collected from the coast of the East Sea in Korea, showed a moderate vasodilatation effect on the basilar arteries of rabbits. Therefore, treatment with farnesylacetones 311 and 312 may
Inhibition of transmethylases by unsaturated carbonyl derivatives.
M Rojas et al.
Die Naturwissenschaften, 67(12), 607-608 (1980-12-01)
Catalytic Epoxypolyene Cyclization via Radicals: A Simple Total Synthesis of Sclareol Oxide and its 8Epimer.
Gansauer A, et al.
Synthesis, 2151-2154 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service