Skip to Content
Merck
All Photos(1)

Key Documents

459984

Sigma-Aldrich

Rhenium

foil, thickness 1.0 mm, 99.98% trace metals basis

Synonym(s):

Rhenium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Re
CAS Number:
Molecular Weight:
186.21
EC Number:
MDL number:
UNSPSC Code:
12141737
PubChem Substance ID:
NACRES:
NA.23

Assay

99.98% trace metals basis

form

foil

description

19.3 μΩ-cm, 20°C

thickness

1.0 mm

bp

5596 °C (lit.)
5627 °C (lit.)

mp

3180 °C (lit.)

density

21.02 g/cm3 (lit.)

SMILES string

[ReH]

InChI

1S/Re

InChI key

WUAPFZMCVAUBPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

13 g = 25 × 25 mm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Dietrich et al.
Inorganic chemistry, 52(3), 1248-1264 (2013-01-15)
New cationic metallo ligands L1-L3 based on bis(terpyridine) ruthenium(II) complexes decorated with differently substituted 2,2'-bipyridines attached via amide groups (5-NHCO-bpy, 4-CONH-bpy, 5-CONH-bpy) were prepared. Coordination of Re(I)Cl(CO)(3) fragments to the bpy unit gives the corresponding bimetallic Ru~Re complexes 1-3. Hydrogen
Cinzia Spagnul et al.
Journal of inorganic biochemistry, 122, 57-65 (2013-03-12)
The synthesis and characterization of two novel water soluble porphyrins with three meso pyridyl rings and one peripheral chelator - either a diethylenetriamine unit (4) or a bipyridyl fragment (8) - for binding to the {(99m)Tc(CO)3}(+) moiety is reported. In
Yanfeng Jiang et al.
Journal of the American Chemical Society, 135(10), 4088-4102 (2013-02-07)
Diiodo Re(I) complexes [ReI2(NO)(PR3)2(L)] (3, L = H2O; 4 , L = H2; R = iPr a, Cy b) were prepared and found to exhibit in the presence of "hydrosilane/B(C6F5)3" co-catalytic systems excellent activities and longevities in the hydrogenation of
Shalina C Bottorff et al.
Inorganic chemistry, 52(6), 2939-2950 (2013-03-06)
The viability of the Huisgen cycloaddition reaction for clickable radiopharmaceutical probes was explored with an alkyne-functionalized 2-[(pyridin-2-ylmethyl)amino]acetic acid (PMAA) ligand system, 3, and fac-[M(I)(OH2)3(CO)3](+) (M = Re, (99m)Tc). Two synthetic strategies, (1) click, then chelate and (2) chelate, then click
Titel Jurca et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(13), 4278-4286 (2013-02-02)
The development of rhenium(I) chemistry has been restricted by the limited structural and electronic variability of the common pseudo-octahedral products fac-[ReX(CO)3L2] (L2 = α-diimine). We address this constraint by first preparing the bidentate bis(imino)pyridine complexes [(2,6-{2,6-Me2C6H3N=CPh}2C5H3N)Re(CO)3X] (X = Cl 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service