Skip to Content
Merck
All Photos(1)

Key Documents

459348

Sigma-Aldrich

Fmoc-Tyr(tBu)-OH

97%

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-tyrosine, Fmoc-O-tert-butyl-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H29NO5
CAS Number:
Molecular Weight:
459.53
Beilstein:
4216652
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D −28°, c = 1 in DMF

mp

153-156 °C (lit.)

SMILES string

CC(C)(C)Oc1ccc(C[C@H](NC(=O)OCC2c3ccccc3-c4ccccc24)C(O)=O)cc1

InChI

1S/C28H29NO5/c1-28(2,3)34-19-14-12-18(13-15-19)16-25(26(30)31)29-27(32)33-17-24-22-10-6-4-8-20(22)21-9-5-7-11-23(21)24/h4-15,24-25H,16-17H2,1-3H3,(H,29,32)(H,30,31)/t25-/m0/s1

InChI key

JAUKCFULLJFBFN-VWLOTQADSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chiao-Ting Yen et al.
European journal of medicinal chemistry, 44(5), 1933-1940 (2008-12-27)
Twenty-four new dipeptide analogs (1-24) of aurantiamide acetate were designed, synthesized, and assayed for effects on superoxide anion generation and elastase release by human neutrophils in response to fMLP/CB. Among them, seven N-(fluorenyl-9-methoxycarbonyl) (Fmoc)-dipeptides (6, 9, 12, 14, 17, 18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service