Skip to Content
Merck
All Photos(1)

Key Documents

458341

Sigma-Aldrich

Poly(1,4-butylene adipate), diol end-capped

average Mn ~1,000

Synonym(s):

1,4-Butanediol-adipic acid copolymer, Adipic acid-1,4-butanediol copolymer, Adipic acid-1,4-butanediol polyester, Poly(1,4-butanediol adipate), Poly(tetramethylene adipate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H[O(CH2)4O2C(CH2)4CO]nCH2CH2CH2CH2OH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~1,000

impurities

1 wt. % 1,4-butanediol

viscosity

350 cP(60 °C)(lit.)

bp

>350 °C (lit.)

mp

50-60 °C (lit.)

hydroxyl functionality

2.0

hydroxyl value

110 mg KOH/g

density

1.08 g/mL at 25 °C (lit.)

SMILES string

OCCCCO.OC(=O)CCCCC(O)=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service