Skip to Content
Merck
All Photos(1)

Key Documents

458147

Sigma-Aldrich

Poly[trimethylolpropane/di(propylene glycol)-alt-adipic acid/phthalic anhydride], polyol

average Mn ~500

Synonym(s):

2-ethyl-2-(hydroxymethyl)-1,3-propanediol, hexanedioic acid and 1,3-isobenzofurandione

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$98.30

$98.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$98.30

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$98.30


Check Cart for Availability

Request a Bulk Order

eq. wt.

200

mol wt

average Mn ~500

refractive index

n20/D 1.472 (lit.)

viscosity

1,500 cP(25 °C)(lit.)

bp

>250 °C (lit.)

mp

<−25 °C (lit.)

hydroxyl functionality

2.5

hydroxyl value

300 mg KOH/g

density

1.11 g/mL at 25 °C (lit.)

SMILES string

O1C(=O)c2c(cccc2)C1=O.O(CC(O)C)CC(O)C.OCC(CO)(CO)CC.OC(=O)CCCCC(=O)O

InChI

1S/C8H4O3.C6H10O4.2C6H14O3/c9-7-5-3-1-2-4-6(5)8(10)11-7;7-5(8)3-1-2-4-6(9)10;1-5(7)3-9-4-6(2)8;1-2-6(3-7,4-8)5-9/h1-4H;1-4H2,(H,7,8)(H,9,10);5-8H,3-4H2,1-2H3;7-9H,2-5H2,1H3

InChI key

OODDSGCGIVJJFS-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service