Skip to Content
Merck
All Photos(1)

Key Documents

456284

Sigma-Aldrich

Fluorene-d10

≥98 atom % D, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$259.00
1 G
$590.00

$259.00


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$259.00
1 G
$590.00

About This Item

Empirical Formula (Hill Notation):
C13D10
CAS Number:
Molecular Weight:
176.28
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$259.00


Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

form

solid

mp

115-118 °C (lit.)

application(s)

electronics

mass shift

M+10

SMILES string

[2H]c1c([2H])c([2H])c2c(c1[2H])-c3c([2H])c([2H])c([2H])c([2H])c3C2([2H])[2H]

InChI

1S/C13H10/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12/h1-8H,9H2/i1D,2D,3D,4D,5D,6D,7D,8D,9D2

InChI key

NIHNNTQXNPWCJQ-QNNBDYQESA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

303.8 °F - closed cup

Flash Point(C)

151.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N D G Chau et al.
Environmental science and pollution research international, 22(12), 9042-9058 (2015-01-13)
Pollution of drinking water sources with agrochemicals is often a major threat to human and ecosystem health in some river deltas, where agricultural production must meet the requirements of national food security or export aspirations. This study was performed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service