Skip to Content
Merck
All Photos(2)

Key Documents

455997

Sigma-Aldrich

Calcium oxalate

≥99.9% trace metals basis

Synonym(s):

Calcium oxalate (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaC2O4
CAS Number:
Molecular Weight:
128.10
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
for analytical purposes
form:
powder and chunks

grade

for analytical purposes

Assay

≥99.9% trace metals basis

form

powder and chunks

impurities

≤1000 ppm Trace Metal Analysis

density

2.2 g/mL at 25 °C (lit.)

SMILES string

O=C1O[Ca]OC1=O

InChI

1S/C2H2O4.Ca/c3-1(4)2(5)6;/h(H,3,4)(H,5,6);/q;+2/p-2

InChI key

QXDMQSPYEZFLGF-UHFFFAOYSA-L

General description

Calcium oxalate (CaC2O4) belongs to the class of oxalates that are associated with soils and lead litter. It forms crystals that are free-living, pathogenic, and plant symbiotic fungi. It is formed by the re-precipitation of calcium as the oxalate.[1]
Purity based on trace metal analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jorge M C Mondego et al.
BMC genomics, 9, 548-548 (2008-11-21)
The basidiomycete fungus Moniliophthora perniciosa is the causal agent of Witches' Broom Disease (WBD) in cacao (Theobroma cacao). It is a hemibiotrophic pathogen that colonizes the apoplast of cacao's meristematic tissues as a biotrophic pathogen, switching to a saprotrophic lifestyle
Atsushi Okada et al.
Clinical and experimental nephrology, 23(5), 710-716 (2019-01-20)
Risk assessment for urinary stones has been mainly based on urinary biochemistry. We attempted to identify the risk factors for urinary stones by statistically analyzing urinary biochemical and inflammation-related factors. Male participants (age, 20-79 years) who visited Nagoya City University Hospital
Jörg Schaller et al.
Scientific reports, 10(1), 2424-2424 (2020-02-14)
More frequent and longer drought periods are predicted threatening agricultural yield. The capacity of soils to hold water is a highly important factor controlling drought stress intensity for plants. Biogenic amorphous silica (bASi) pools in soils are in the range
Masao Tsujihata
International journal of urology : official journal of the Japanese Urological Association, 15(2), 115-120 (2008-02-14)
Formation of calcium oxalate stones tends to increase with age and begins from the attachment of a crystal formed in the cavity of renal tubules to the surface of renal tubular epithelial cells. Though most of the crystals formed in
Philipp Schlee et al.
Chemical science, 10(10), 2980-2988 (2019-04-19)
We have discovered a very simple method to address the challenge associated with the low volumetric energy density of free-standing carbon nanofiber electrodes for supercapacitors by electrospinning Kraft lignin in the presence of an oxidizing salt (NaNO3) and subsequent carbonization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service