Skip to Content
Merck
All Photos(1)

Key Documents

455024

Sigma-Aldrich

Poly(propylene glycol) diacrylate

average Mn ~800, contains 100 ppm BHT as inhibitor, 100 ppm MEHQ as inhibitor

Synonym(s):

Poly(propylene oxide) diacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$82.70

$82.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$82.70

About This Item

Linear Formula:
H2C=CHCO(OC3H6)nO2CCH=CH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$82.70


Check Cart for Availability

Request a Bulk Order

mol wt

average Mn ~800

Quality Level

contains

100 ppm BHT as inhibitor
100 ppm MEHQ as inhibitor

refractive index

n20/D 1.452

density

1.01 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

OCCCO.OC(=O)C=C

InChI

1S/C3H4O2.C3H8O2/c1-2-3(4)5;4-2-1-3-5/h2H,1H2,(H,4,5);4-5H,1-3H2

InChI key

TXHCDEYHZPHRAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chueh-Yu Wu et al.
Science advances, 6(45) (2020-11-06)
The ability to create uniform subnanoliter compartments using microfluidic control has enabled new approaches for analysis of single cells and molecules. However, specialized instruments or expertise has been required, slowing the adoption of these cutting-edge applications. Here, we show that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service