Skip to Content
Merck
All Photos(2)

Documents

454907

Sigma-Aldrich

Cellulose propionate

average Mw ~130,000, average Mn ~70,000

Synonym(s):

Cellulose propionate, Propionyl cellulose

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

mol wt

average Mn ~70,000
average Mw ~130,000

contains

7 wt. % plasticizer

impact strength

203 J/m (Izod, 23 °C, notched)

hardness

95 (Rockwell R, ASTM D 785)

transition temp

flow temperature 347 °F

density

1.22 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)OC[C@H]1O[C@@H](OC(=O)CC)[C@H](OC(=O)CC)[C@@H](OC(=O)CC)[C@@H]1O[C@@H]2O[C@H](COC(=O)CC)[C@@H](OC(=O)CC)[C@H](OC(=O)CC)[C@H]2OC(=O)CC

InChI

1S/C36H54O19/c1-9-21(37)45-17-19-29(49-23(39)11-3)31(50-24(40)12-4)34(53-27(43)15-7)36(48-19)55-30-20(18-46-22(38)10-2)47-35(54-28(44)16-8)33(52-26(42)14-6)32(30)51-25(41)13-5/h19-20,29-36H,9-18H2,1-8H3/t19-,20-,29-,30-,31+,32+,33-,34-,35+,36+/m1/s1

InChI key

DQEFEBPAPFSJLV-WLTGXWPBSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service