Skip to Content
Merck
All Photos(1)

Documents

454273

Sigma-Aldrich

5-Ethyl-5-methylhydantoin

97%

Synonym(s):

5-Ethyl-5-methyl-2,4-imidazolidinedione, NSC 1020

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N2O2
CAS Number:
Molecular Weight:
142.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

144-150 °C (lit.)

SMILES string

CCC1(C)NC(=O)NC1=O

InChI

1S/C6H10N2O2/c1-3-6(2)4(9)7-5(10)8-6/h3H2,1-2H3,(H2,7,8,9,10)

InChI key

VSJRBQDMBFFHMC-UHFFFAOYSA-N

Application

Reactant for synthesis of nociceptin/orphanin FQ analogues

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preferential crystallisation and comparative crystal growth study between pure enantiomer and racemic mixture of a chiral molecule: 5-ethyl-5-methylhydantoin.
Beilles S, et al.
Chemical Engineering Science, 56(7), 2281-2294 (2001)
Palash K Sarker et al.
International journal of molecular sciences, 13(1), 1006-1017 (2012-02-09)
Aqueous solutions of isovaline and its precursor molecule, 5-ethyl-5-methylhydantoin, were irradiated with ultraviolet and γ-ray photons, to evaluate their structural stability against space radiation. The degree of photolysis was measured and irradiation products were identified using chiral, reversed-phase and ion-exchange

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service