Skip to Content
Merck
All Photos(3)

Key Documents

453188

Sigma-Aldrich

D-Glucose-1,2-13C2

99 atom % 13C

Synonym(s):

Labeled Glucose, Dextrose-1,2-13C2

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$417.00
500 MG
$1,060.00
1 G
$1,860.00

$417.00


Available to ship on17 April 2025Details


Request more information

Select a Size

Change View
100 MG
$417.00
500 MG
$1,060.00
1 G
$1,860.00

About This Item

Empirical Formula (Hill Notation):
13C2C4H12O6
CAS Number:
Molecular Weight:
182.14
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

$417.00


Available to ship on17 April 2025Details


Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

99% (CP)

form

powder

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

mp

150-152 °C (lit.)

mass shift

M+2

storage temp.

room temp

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[13C@@H](O)[13CH]=O

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6+/m0/s1/i1+1,3+1

InChI key

GZCGUPFRVQAUEE-SZEFTZBMSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

D-Glucose-1,2-13C2 can be used:
  • As a stable isotope in the in vivo study of dynamic metabolic profiling of pancreatic tumor.[1]
  • To study the nonoxidative pentose phosphate pathway using a single-tracer method.[2]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[1, 2-13C2]-D-glucose profiles of the serum, liver, pancreas, and DMBA-induced pancreatic tumors of rats
Boros L, et al.
Pancreas, 31(4), 337-343 (2005)
Mass isotopomer study of the nonoxidative pathways of the pentose cycle with [1, 2-13C2] glucose
Lee, WP, et al.
American Journal of Physiology. Endocrinology and Metabolism, 274(5), E843-E851 (1998)
Joanna L Gillis et al.
eLife, 10 (2021-08-13)
Alterations to the androgen receptor (AR) signalling axis and cellular metabolism are hallmarks of prostate cancer. This study provides insight into both hallmarks by uncovering a novel link between AR and the pentose phosphate pathway (PPP). Specifically, we identify 6-phosphogluoconate

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service