Skip to Content
Merck
All Photos(1)

Documents

450898

Sigma-Aldrich

Gallium(III) chloride

anhydrous, beads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Gallium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaCl3
CAS Number:
Molecular Weight:
176.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

vapor pressure

1 mmHg ( 48 °C)

Assay

99.99% trace metals basis

form

beads

reaction suitability

reagent type: catalyst
core: gallium

impurities

≤150.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

78 °C (lit.)

density

2.47 g/mL at 25 °C (lit.)

SMILES string

Cl[Ga](Cl)Cl

InChI

1S/3ClH.Ga/h3*1H;/q;;;+3/p-3

InChI key

UPWPDUACHOATKO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for a number of transformations such as the ring opening and recyclization of epoxides or the trimerization of alkynes.

Features and Benefits

Lower-temperature solvothermal techniques using GaCl3 and NaN3 or Li3N result in nanocrystalline GaN.
Radical mediator and catalyst in a number of organic bond formation reactions.

Packaging

product is packed in vials

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tiina Ujula et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 12(3), 259-268 (2009-10-03)
The aim of the study was to compare (68)Ga-chloride with 2-[(18)F]fluoro-2-deoxy-D: -glucose (FDG) for the imaging of pancreatic xenografts. Rats with subcutaneous human pancreatic adenocarcinoma xenografts were evaluated in vivo by dynamic positron emission tomography (PET) and ex vivo by
Han-Gook Cho
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(7), 1517-1528 (2003-04-26)
Remarkably large blue shifts of the nu2 C [triple bond] N stretch, nu4 C-C stretch, and nu8 CCN deformation bands of CD3CN are observed in the infrared and Raman spectra of CD3CN solution of GaCl3, resulting from the donor-acceptor interaction
Amir R Jalilian et al.
Acta pharmaceutica (Zagreb, Croatia), 59(4), 421-429 (2009-11-19)
beta-Human chorionic gonadotropin (beta-hCG) was successively labeled with [(67)Ga]-gallium chloride after conjugation with freshly prepared diethylenetriaminepentaacetic acid dianhydride (ccDTPA). After solid phase purification of the radiolabeled hormone, high performance liquid chromatography showed radiochemical purity higher than 95 % under optimized
Karin Grijm et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 46(9), 1421-1426 (2005-09-15)
The prognosis in some forms of idiopathic interstitial pneumonia (IIP), especially idiopathic pulmonary fibrosis (IPF) and fibrotic nonspecific interstitial pneumonia (NSIP), is still poor. A minority of patients will respond to immunosuppressive treatment. In patients with IPF or fibrotic NSIP
C Caraco et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 49(12), 1477-1479 (1998-09-24)
Previous studies report that deferoxamine (DFO) binds metallic ions such as Fe3+, In3+ and Ga3+ with very high affinity. This property of DFO has been utilized to label DFO-coupled compounds with radiometals such as 67Ga and 111In. We have studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service