Skip to Content
Merck
All Photos(2)

Key Documents

450480

Sigma-Aldrich

4,4′-(1,3-Phenylenediisopropylidene)bisaniline

98%

Synonym(s):

Bisaniline M

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$175.00

$175.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$175.00

About This Item

Linear Formula:
C6H4[C(CH3)2C6H4NH2]2
CAS Number:
Molecular Weight:
344.49
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$175.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

110-114 °C (lit.)

SMILES string

CC(C)(c1ccc(N)cc1)c2cccc(c2)C(C)(C)c3ccc(N)cc3

InChI

1S/C24H28N2/c1-23(2,17-8-12-21(25)13-9-17)19-6-5-7-20(16-19)24(3,4)18-10-14-22(26)15-11-18/h5-16H,25-26H2,1-4H3

InChI key

KWOIWTRRPFHBSI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service