Skip to Content
Merck
All Photos(2)

Key Documents

450308

Sigma-Aldrich

Aluminum sulfate hydrate

99.99% trace metals basis

Synonym(s):

Aluminum sulfate hydrate, Dialuminum trisulfate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$97.10

$97.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$97.10

About This Item

Linear Formula:
Al2(SO4)3 · xH2O
CAS Number:
Molecular Weight:
342.15 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder and chunks

$97.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99.99% trace metals basis

form

powder and chunks

impurities

≤30.0 ppm Trace Metal Analysis

density

1.69 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Al+3].[Al+3].[H]O[H].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Al.3H2O4S.H2O/c;;3*1-5(2,3)4;/h;;3*(H2,1,2,3,4);1H2/q2*+3;;;;/p-6

InChI key

BUACSMWVFUNQET-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service