Skip to Content
Merck
All Photos(1)

Key Documents

450014

Sigma-Aldrich

Sodium chloride

AnhydroBeads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
99.99% trace metals basis
solubility:
water: soluble (358 g/l at 20 °C)

vapor pressure

1.33 (1.00 mmHg) at 865 °C (1589 °F) hPa

product line

AnhydroBeads

Assay

99.99% trace metals basis

particle size

−10 mesh

mp

801 °C (lit.)

solubility

water: soluble (358 g/l at 20 °C)

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Facilitates the cross-coupling of organostannanes with iodides without using palladium.[1]

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copper- and Manganese-Catalyzed Cross-Coupling of Organostannanes with Organic Iodides in the Presence of Sodium Chloride.
Suk-Ku Kang et al.
The Journal of organic chemistry, 62(13), 4208-4209 (1997-06-27)
Zlatko Kopecki et al.
Oncotarget, 6(34), 36426-36440 (2015-10-27)
Flightless I (Flii) is an actin remodeling protein that affects cellular processes including adhesion, proliferation and migration. In order to determine the role of Flii during carcinogenesis, squamous cell carcinomas (SCCs) were induced in Flii heterozygous (Flii+/-), wild-type and Flii
Ram S Puranam et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(23), 8866-8881 (2015-06-13)
We identified a family in which a translocation between chromosomes X and 14 was associated with cognitive impairment and a complex genetic disorder termed "Genetic Epilepsy and Febrile Seizures Plus" (GEFS(+)). We demonstrate that the breakpoint on the X chromosome
Qian Chen et al.
Genes & development, 29(12), 1285-1297 (2015-06-26)
The Hippo signaling pathway converges on YAP to regulate growth, differentiation, and regeneration. Previous studies with overexpressed proteins have shown that YAP is phosphorylated by its upstream kinase, Lats1/2, on multiple sites, including an evolutionarily conserved 14-3-3-binding site whose phosphorylation
Viktor Posse et al.
Nucleic acids research, 43(5), 2615-2624 (2015-02-19)
A single-subunit RNA polymerase, POLRMT, transcribes the mitochondrial genome in human cells. Recently, a factor termed as the mitochondrial transcription elongation factor, TEFM, was shown to stimulate transcription elongation in vivo, but its effect in vitro was relatively modest. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service