Skip to Content
Merck
All Photos(1)

Key Documents

449423

Sigma-Aldrich

2-Chlorophenol-3,4,5,6-d4

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$260.00
250 MG
$667.00

$260.00


Available to ship on22 April 2025Details


Request more information

Select a Size

Change View
100 MG
$260.00
250 MG
$667.00

About This Item

Linear Formula:
ClC6D4OH
CAS Number:
Molecular Weight:
132.58
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$260.00


Available to ship on22 April 2025Details


Request more information

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.558 (lit.)

bp

175-176 °C (lit.)

mp

8 °C (lit.)

density

1.279 g/mL at 25 °C

mass shift

M+4

SMILES string

[2H]c1c([2H])c([2H])c(Cl)c(O)c1[2H]

InChI

1S/C6H5ClO/c7-5-3-1-2-4-6(5)8/h1-4,8H/i1D,2D,3D,4D

InChI key

ISPYQTSUDJAMAB-RHQRLBAQSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service