Skip to Content
Merck
All Photos(1)

Documents

449067

Sigma-Aldrich

4-(Hexyloxy)benzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5OC6H4CHO
CAS Number:
Molecular Weight:
206.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.529 (lit.)

bp

154-155 °C/6 mmHg (lit.)

density

0.992 g/mL at 25 °C (lit.)

SMILES string

CCCCCCOc1ccc(C=O)cc1

InChI

1S/C13H18O2/c1-2-3-4-5-10-15-13-8-6-12(11-14)7-9-13/h6-9,11H,2-5,10H2,1H3

InChI key

GWXUVWKBVROFDM-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescence and FTIR Spectra Analysis of Trans-A2B2-Substituted Di-and Tetra-Phenyl Porphyrins.
Sen P, et al.
Materials, 3(8), 4446-4475 (2010)
Soluble phenyl-or alkoxyphenyl-substituted rigid-rod polyamides and polyimides containing m-terphenyls in the main chain.
Spiliopoulos LK and Mikroyannidis JA.
Macromolecules, 31(4), 1236-1245 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service