Skip to Content
Merck
All Photos(1)

Key Documents

448664

Sigma-Aldrich

[3-(Diethylamino)propyl]trimethoxysilane

96%

Synonym(s):

N,N-Diethyl-3-(trimethoxysilyl)propylamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$99.80

$99.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$99.80

About This Item

Linear Formula:
(C2H5)2N(CH2)3Si(OCH3)3
CAS Number:
Molecular Weight:
235.40
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$99.80


Check Cart for Availability

Request a Bulk Order

Assay

96%

refractive index

n20/D 1.423 (lit.)

bp

120 °C/20 mmHg (lit.)

density

0.95 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCN(CC)CCC[Si](OC)(OC)OC

InChI

1S/C10H25NO3Si/c1-6-11(7-2)9-8-10-15(12-3,13-4)14-5/h6-10H2,1-5H3

InChI key

ZLDHYRXZZNDOKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

[3-(Diethylamino)propyl]trimethoxysilane was covalently bonded to mesoporous titania–silica mixed oxides to act as epoxidation catalysts.{55} [3-(Diethylamino)propyl]trimethoxysilane, immobilized on mesoporous silicas (SBA-15) may be used as adsorbents to capture CO2. {56}

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Primary, secondary, and tertiary amines for CO2 capture: Designing for mesoporous CO2 adsorbents
Ko YG, et al.
Journal of Colloid and Interface Science, 361(2), 594-602 (2011)
Amine-modified titania?silica hybrid gels as epoxidation catalysts
Muller CA, et al.
Applied Catalysis A: General, 201(2), 253-261 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service