Skip to Content
Merck
All Photos(1)

Key Documents

448257

Sigma-Aldrich

5-(Methylthio)-1H-tetrazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4N4S
CAS Number:
Molecular Weight:
116.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

149-152 °C (lit.)

SMILES string

CSc1nnn[nH]1

InChI

1S/C2H4N4S/c1-7-2-3-5-6-4-2/h1H3,(H,3,4,5,6)

InChI key

ZBXNFTFKKOSPLD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nabakumar Pramanik et al.
International journal of biomaterials, 2009, 512417-512417 (2010-02-05)
A novel bioanalogue hydroxyapatite (HAp)/chitosan phosphate (CSP) nanocomposite has been synthesized by a solution-based chemical methodology with varying HAp contents from 10 to 60% (w/w). The interfacial bonding interaction between HAp and CSP has been investigated through Fourier transform infrared
W Koźmiński et al.
Solid state nuclear magnetic resonance, 4(2), 121-124 (1995-02-01)
15N and 13C cross-polarization magic-angle spinning nuclear magnetic resonance (CP-MAS NMR) spectra of 5-thiomethyltetrazole have been measured. Two kinds of tetrazole molecules are found in the hydrogen bond chain system depending on the configuration of the S-CH3 group in relation
B Guan et al.
Acta endocrinologica (Bucharest, Romania : 2005), 15(2), 158-164 (2019-09-12)
Endoplasmic reticulum stress (ERS) is suspected as an important factor in the initiation of insulin resistance. To explore the effects of exendin-4 (Ex-4) on the endoplasmic reticulum stress (ERS)-mediated insulin resistance in 3T3-L1 adipocytes. In our study, 3T3-L1 adipocytes were
Laura Tapella et al.
Pituitary, 20(3), 311-318 (2016-11-18)
Endocrine disruptors are known to modulate a variety of endocrine functions and increase the risk for neoplasia. Epidemiological data reported increased prevalence of pituitary tumors in high industrial areas while genotyping studies showed that mutations in the aryl hydrocarbon receptor
The Effectivity of 1H-Triazoles and-Tetrazoles as Activators in Acid-Catalyzed Phosphoramidite Alcoholysis.
Nurminen E, et al.
Helvetica Chimica Acta, 86(6), 2005-2008 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service