Skip to Content
Merck
All Photos(2)

Key Documents

448109

Sigma-Aldrich

2-Bromo-3-methylpyridine

95%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$161.00
25 ML
$295.00

$161.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$161.00
25 ML
$295.00

About This Item

Empirical Formula (Hill Notation):
C6H6BrN
CAS Number:
Molecular Weight:
172.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$161.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

refractive index

n20/D 1.568 (lit.)

bp

218-219 °C (lit.)

density

1.544 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Cc1cccnc1Br

InChI

1S/C6H6BrN/c1-5-3-2-4-8-6(5)7/h2-4H,1H3

InChI key

PZSISEFPCYMBDL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 2, 2'-Bipyridines: Versatile Building Blocks for Sexy Architectures and Functional Nanomaterials.
European Journal of Organic Chemistry, 2, 235-254 (2004)
Copper (ii) complexes of 2-halo-3-methylpyridine: synthesis, structure, and magnetic behaviour of Cu (2-X-3-CH 3 py) 2 X' 2 [X, X'= chlorine or bromine; py= pyridine].
Herringer SN, et al.
Dalton Transactions, 40(16), 4242-4252 (2011)
Lynne H Thomas et al.
Acta crystallographica. Section C, Crystal structure communications, 69(Pt 11), 1279-1288 (2013-11-07)
Controlled introduction of proton transfer into the design of a series of molecular complexes is described, delivering the systematic production of ionic molecular complexes (molecular salts). The controlled production of molecular salts has relevance as a potential strategy in the
Kelnner W R de França et al.
The Journal of organic chemistry, 67(6), 1838-1842 (2002-03-16)
2,2'-Bipyridine (bpy) and a series of dimethyl-2,2'-bipyridines were synthesized from 2-bromopyridine and 2-bromomethylpyridines, respectively, using an electrochemical process catalyzed by nickel complexes. The method is simple and efficient, with isolated yields between 58 and 98% according to the structure. We
Alan C Spivey et al.
Organic letters, 9(5), 891-894 (2007-02-10)
[reaction: see text] The scope and limitations of the conjugate addition of 2- and the first 4-pyridyl Gilman homocuprates to various alpha,beta-unsaturated Michael acceptors are delineated. The conjugate addition of the cuprate of 2-bromo-3-methylpyridine to (E)-methyl crotonate then diastereoselective enolate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service