447048
1,2-Bis(trichlorosilyl)ethane
97%
Synonym(s):
1,1,1,4,4,4-Hexachlorodisilethylene
About This Item
Recommended Products
Assay
97%
form
solid
refractive index
n20/D 1.475 (lit.)
bp
202 °C (lit.)
mp
27-29 °C (lit.)
density
1.483 g/mL at 25 °C (lit.)
SMILES string
Cl[Si](Cl)(Cl)CC[Si](Cl)(Cl)Cl
InChI
1S/C2H4Cl6Si2/c3-9(4,5)1-2-10(6,7)8/h1-2H2
InChI key
WDVUXWDZTPZIIE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
No data available
Flash Point(C)
No data available
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service