Skip to Content
Merck
All Photos(2)

Key Documents

446289

Sigma-Aldrich

Trichloro(pentamethylcyclopentadienyl)titanium(IV)

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$493.00

$493.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$493.00

About This Item

Empirical Formula (Hill Notation):
C10H15Cl3Ti
CAS Number:
Molecular Weight:
289.45
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$493.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

reaction suitability

core: titanium
reagent type: catalyst

mp

200 °C (dec.) (lit.)

SMILES string

Cl[Ti](Cl)Cl.C[C]1[C](C)[C](C)[C](C)[C]1C

InChI

1S/C10H15.3ClH.Ti/c1-6-7(2)9(4)10(5)8(6)3;;;;/h1-5H3;3*1H;/q;;;;+3/p-3

InChI key

QCEOZLISXJGWSW-UHFFFAOYSA-K

Application

Catalyst for:
  • Transesterification reactions
  • Styrene polymerization

  • Piano-stool catalyst

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service