Skip to Content
Merck
All Photos(1)

Key Documents

445487

Sigma-Aldrich

Ethyl N,N-dimethyloxamate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCOCO2C2H5
CAS Number:
Molecular Weight:
145.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.442 (lit.)

bp

105-106 °C/7 mmHg (lit.)

density

1.068 g/mL at 25 °C (lit.)

functional group

amide
ester

SMILES string

CCOC(=O)C(=O)N(C)C

InChI

1S/C6H11NO3/c1-4-10-6(9)5(8)7(2)3/h4H2,1-3H3

InChI key

HMALWDVRMHVUAW-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ab initio calculations of the gas-phase elimination kinetics of ethyl oxamate, ethyl oxanilate, and ethyl N, N-dimethyl oxamate.
Mora JR, et al.
Journal of Physical Organic Chemistry, 19(8-9), 503-511 (2006)
Kinetics of the homogeneous, unimolecular elimination reactions of ethyl oxamate, ethyl N,N-dimethyloxamate and ethyl oxanilate in the gas phase.
Chacin EV, et al.
Journal of Physical Organic Chemistry, 18(6), 539-545 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service