Skip to Content
Merck
All Photos(1)

Key Documents

44493

Sigma-Aldrich

1,3-Bis(3-cyanopropyl)imidazolium bis(trifluoromethylsulfonyl)imide

≥98.0% (HPLC/T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15F6N5O4S2
CAS Number:
Molecular Weight:
483.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (HPLC/T)

impurities

≤0.1% water
≤10 mg/kg halogenes (as chloride)

refractive index

n20/D 1.456

SMILES string

FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.N#CCCCn1cc[n+](CCCC#N)c1

Application

Nitrile-functionalized ionic liquid showing superior characteristics in Suzuki and Stille coupling reactions[1].

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongbin Zhao et al.
Journal of the American Chemical Society, 126(48), 15876-15882 (2004-12-02)
A series of relatively low-cost ionic liquids, based on the N-butyronitrile pyridinium cation [C(3)CNpy](+), designed to improve catalyst retention, have been prepared and evaluated in Suzuki and Stille coupling reactions. Depending on the nature of the anion, these salts react

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service