Skip to Content
Merck
All Photos(1)

Key Documents

444774

Sigma-Aldrich

(4-Bromophenoxy)-tert-butyldimethylsilane

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$142.00
25 ML
$376.00

$142.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$142.00
25 ML
$376.00

About This Item

Linear Formula:
BrC6H4OSi(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
287.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$142.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.511 (lit.)

bp

137 °C/25 mmHg (lit.)

density

1.174 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

CC(C)(C)[Si](C)(C)Oc1ccc(Br)cc1

InChI

1S/C12H19BrOSi/c1-12(2,3)15(4,5)14-11-8-6-10(13)7-9-11/h6-9H,1-5H3

InChI key

DLGZGLKSNRKLSM-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Hee Ahn et al.
Bioorganic & medicinal chemistry letters, 12(15), 1941-1946 (2002-07-13)
A new series of 1,2-naphthoquinone derivatives was synthesized by various synthetic methods and evaluated for their ability to inhibit protein tyrosine phosphatase 1B (PTP1B). 1,2-Naphthoquinone derivatives with substituent at R(4) position showed submicromolar inhibitory activity, and compound 24 demonstrated 10-

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service