Skip to Content
Merck
All Photos(1)

Key Documents

444235

Sigma-Aldrich

(S)-(−)-α-Hydroxy-γ-butyrolactone

97%, optical purity ee: 96% (GLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

optical activity

[α]23/D −68°, c = 1.15 in chloroform

optical purity

ee: 96% (GLC)

refractive index

n20/D 1.467 (lit.)

bp

133 °C/10 mmHg (lit.)

density

1.309 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O[C@H]1CCOC1=O

InChI

1S/C4H6O3/c5-3-1-2-7-4(3)6/h3,5H,1-2H2/t3-/m0/s1

InChI key

FWIBCWKHNZBDLS-VKHMYHEASA-N

Application

Used to prepare the polyether antibiotic monensin,[1] functionalized D-ring side chains of vitamin D analogs,[2] and pesticides.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui B Sun et al.
Connective tissue research, 53(2), 180-186 (2011-12-14)
Joint loading is a recently developed loading modality, which can enhance bone formation and accelerate healing of bone fracture. Since mechanical stimulation alters expression of matrix metalloproteinases (MMPs) in chondrocytes, a question addressed herein was, does joint loading alter actions
Buser, H.P. et al.
Tetrahedron, 47, 5709-5709 (1991)
Collum, D.B. et al.
Journal of the American Chemical Society, 102, 2118-2118 (1980)
S.-J. Shiuey et al.
The Journal of Organic Chemistry, 53, 1040-1040 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service