Skip to Content
Merck
All Photos(1)

Key Documents

44162

Sigma-Aldrich

1,3-Bis[bis(2-pyridylmethyl)amino]-2-propanol solution

≥97.0% (HPLC), 10% in isopropanol

Synonym(s):

2-Hydroxy-N,N,N′,N′-tetrakis(2-pyridylmethyl)-1,3-propanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30N6O
CAS Number:
Molecular Weight:
454.57
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (HPLC)

concentration

10% in isopropanol

density

0.817 g/mL at 20 °C (lit.)

SMILES string

OC(CN(Cc1ccccn1)Cc2ccccn2)CN(Cc3ccccn3)Cc4ccccn4

Other Notes

Ligand forming dinuclear metal complexes for efficient RNA hydrolysis; structure of zinc complexes

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Adams et al.
J. Chem. Soc., Dalton Trans., 925-925 (2002)
M Yashiro et al.
Journal of biochemistry, 120(6), 1067-1069 (1996-12-01)
A dinuclear La3+ complex hydrolyzes the phosphodiester bond in diribonucleotides efficiently under mild conditions. The rate of hydrolysis is remarkably accelerated on the complex formation between La3+ and TPHP (TPHP = N,N,N',N'-tetrakis [(2-pyridyl)methyl]-2-hydroxy-1,3-diaminopropane), and the highest activity is attained when
A Ishikubo et al.
Nucleic acids symposium series, 34(34), 85-86 (1995-01-01)
Dinuclear Zn(II) and La(III) complexes with TPHP efficiently hydrolyze a dinucleotide, ApA, under mild conditions (TPHP = N, N, N', N'-tetrakis[(2-pyridyl)-methyl]-2-hydroxy-1,3-diaminopropane). [Zn2(TPHP)]3+ hydrolyzes ApA with an extremely high activity; The pseudo-first-order rate constant is 8.4 x 10(-4) h-1 at pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service