Skip to Content
Merck
All Photos(1)

Key Documents

440442

Sigma-Aldrich

O-(Trimethylsilyl)hydroxylamine

technical grade, 90%

Synonym(s):

Aminoxytrimethylsilane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$109.00
5 G
$247.00

$109.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$109.00
5 G
$247.00

About This Item

Linear Formula:
(CH3)3SiONH2
CAS Number:
Molecular Weight:
105.21
Beilstein:
2035893
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$109.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.405 (lit.)

bp

98-100 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)ON

InChI

1S/C3H11NOSi/c1-6(2,3)5-4/h4H2,1-3H3

InChI key

AEKHNNJSMVVESS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

O-(Trimethylsilyl)hydroxylamine is an O-substituted hydroxylamine. It is formed as an intermediate during the trimethylsilyl chloride catalyzed synthesis of α-branched amines.[1]

Application

O-(Trimethylsilyl)hydroxylamine may be used for the preparation of O-trimethylsilyl oxime ethers.[2]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemoselective addition of organometallics to oxime ethers.
Tavakol H,et al.
Journal of Organometallic Chemistry, 692(10), 1924-1927 (2007)
Ying Fu et al.
Organic & biomolecular chemistry, 10(38), 7669-7672 (2012-08-14)
A general procedure for the nucleophilic addition of organozinc halides with nitrones in the presence of trimethylsilyl chloride has been developed. Trimethylsilyl chloride was found to be both an indispensable reaction promoter and a ready hydroxylamine protective agent in these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service