Skip to Content
Merck
All Photos(1)

Key Documents

440329

Sigma-Aldrich

Butyl (S)-(−)-2-pyrrolidone-5-carboxylate

98%

Synonym(s):

Butyl L-pyroglutamate

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$55.90

$55.90


Only 0 left in stock

Request a Bulk Order

Select a Size

Change View
5 ML
$55.90

About This Item

Empirical Formula (Hill Notation):
C9H15NO3
CAS Number:
Molecular Weight:
185.22
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$55.90


Only 0 left in stock

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

optical activity

[α]20/D −13°, neat

refractive index

n20/D 1.403 (lit.)

bp

180 °C/6.5 mmHg (lit.)

density

1.104 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCOC(=O)[C@@H]1CCC(=O)N1

InChI

1S/C9H15NO3/c1-2-3-6-13-9(12)7-4-5-8(11)10-7/h7H,2-6H2,1H3,(H,10,11)/t7-/m0/s1

InChI key

QBBAJUQOYIURHA-ZETCQYMHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service