Skip to Content
Merck
All Photos(1)

Key Documents

437751

Sigma-Aldrich

Phenethyl isobutyrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2CH2CH2C6H5
CAS Number:
Molecular Weight:
192.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

refractive index

n20/D 1.4873 (lit.)

bp

250 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

SMILES string

CC(C)C(=O)OCCc1ccccc1

InChI

1S/C12H16O2/c1-10(2)12(13)14-9-8-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3

InChI key

JDQVBGQWADMTAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenethyl isobutyrate belongs to the fragrance structural group, Aryl Alkyl Alcohol Simple Acid Esters (AAASAE). It is reported as a fragrance ingredient. Toxicologic and dermatologic review on phenethyl isobutyrate has been reported.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D McGinty et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50 Suppl 2, S416-S419 (2012-03-13)
A toxicologic and dermatologic review of phenethyl isobutyrate when used as a fragrance ingredient is presented. Phenethyl isobutyrate is a member of the fragrance structural group Aryl Alkyl Alcohol Simple Acid Esters (AAASAE). The AAASAE fragrance ingredients are prepared by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service