Skip to Content
Merck
All Photos(1)

Key Documents

436941

Sigma-Aldrich

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate

97%

Synonym(s):

EMIM Otf

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11F3N2O3S
CAS Number:
Molecular Weight:
260.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.435 (lit.)

bp

>350 °C (lit.)

density

1.387 g/mL at 25 °C (lit.)

SMILES string

CCn1cc[n+](C)c1.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/C6H11N2.CHF3O3S/c1-3-8-5-4-7(2)6-8;2-1(3,4)8(5,6)7/h4-6H,3H2,1-2H3;(H,5,6,7)/q+1;/p-1

InChI key

ZPTRYWVRCNOTAS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Recently used in the preparation of hydrophobic, highly conductive ambient-temperature molten salts, and in the preparation of dual intercalating molten electrolyte batteries.

replaced by

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

425.3 °F - closed cup

Flash Point(C)

218.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Bonhôte et al.
Inorganic chemistry, 35(5), 1168-1178 (1996-02-28)
New, hydrophobic ionic liquids with low melting points (<-30 degrees C to ambient temperature) have been synthesized and investigated, based on 1,3-dialkyl imidazolium cations and hydrophobic anions. Other imidazolium molten salts with hydrophilic anions and thus water-soluble are also described.
Journal of the Electrochemical Society, 141, L73-L73 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service