Skip to Content
Merck
All Photos(1)

Key Documents

435880

Sigma-Aldrich

Lead(II) iodate

99.99% trace metals basis

Synonym(s):

Lead diiodate, Lead iodate (Pb(IO3 )2 )

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(IO3)2
CAS Number:
Molecular Weight:
557.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.99% trace metals basis

form

powder

mp

300 °C (dec.) (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[PbH2++].[O-]I(=O)=O.[O-]I(=O)=O

InChI

1S/2HIO3.Pb/c2*2-1(3)4;/h2*(H,2,3,4);/q;;+2/p-2

InChI key

DRHWBADNSVQEGH-UHFFFAOYSA-L

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Ox. Sol. 3 - Repr. 1A - STOT RE 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allen E Haddrell et al.
The journal of physical chemistry. A, 116(40), 9941-9953 (2012-09-18)
The most used instrument in single particle hygroscopic analysis over the past thirty years has been the electrodynamic balance (EDB). Two general assumptions are made in hygroscopic studies involving the EDB. First, it is assumed that the net charge on
Barbara Krassowska-Swiebocka et al.
Molecules (Basel, Switzerland), 10(2), 394-400 (2007-11-17)
Benzene, halobenzenes, and a number of more or less deactivated arenes,including nitrobenzene, readily reacted in anhydrous HIO3/AcOH/Ac2O/conc. H2SO4 mixtures to probably give ArIO2 intermediates or other hypervalent species (not isolated). The final reaction mixtures were poured into excess aq. Na2SO3
K J Hill et al.
Biochimica et biophysica acta, 1326(1), 37-46 (1997-05-22)
Liposomes were prepared from phospholipid mixtures of dipalmitoylphosphatidylcholine (DPPC) and phosphatidylinositol (PI), encapsulating the enzymes glucose oxidase (GO) and GO in combination with horse radish peroxidase (HRP) by both extrusion (VET) and reverse-phase evaporation (REV). The optimum level of PI
Colour reactions of iodic acid as reagent for identifying drugs by thin-layer chromatography.
G Cavazzutti et al.
Journal of chromatography, 268(3), 528-534 (1983-10-07)
Amanda C Bean et al.
Inorganic chemistry, 44(15), 5209-5211 (2005-07-19)
The hydrothermal reaction of a (239)Pu(IV) stock solution in the presence of iodic acid and 1 M KOH produces reddish-brown single crystals of (PuO(2))(2)(IO(3))(OH)(3). The structure consists of two-dimensional layers forming in the ac plane and is the first single-crystal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service