Skip to Content
Merck
All Photos(1)

Key Documents

435872

Sigma-Aldrich

Lead(II) niobate

−200 mesh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(NbO3)2
CAS Number:
Molecular Weight:
489.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Pricing and availability is not currently available. Contact Technical Service

particle size

−200 mesh

SMILES string

O=[Nb](=O)O[PbH2]O[Nb](=O)=O

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service