Skip to Content
Merck
All Photos(1)

Key Documents

435732

Sigma-Aldrich

5-(tert-Butyldimethylsilyloxy)-1-pentanol

90%

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$512.00

$512.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$512.00

About This Item

Linear Formula:
(CH3)3CSi(CH3)2O(CH2)5OH
CAS Number:
Molecular Weight:
218.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$512.00


Check Cart for Availability

Request a Bulk Order

Assay

90%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

140 °C/6 mmHg (lit.)

density

0.885 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

C[Si](C)(OCCCCCO)C(C)(C)C

InChI

1S/C11H26O2Si/c1-11(2,3)14(4,5)13-10-8-6-7-9-12/h12H,6-10H2,1-5H3

InChI key

ZPADIAOSJBISTI-UHFFFAOYSA-N

General description

5-(tert-Butyldimethylsilyloxy)-1-pentanol participates in the synthesis of well-defined mono- and ditelechelic polyphosphazenes.[1]

Other Notes

remainder 1,5-bis(tert-butyldimethylsilyoxy)pentane

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Telechelic polyphosphazenes: Reaction of living poly (dichlorophosphazene) chains with alkoxy and aryloxy phosphoranimines.
Allcock HR, et al.
Macromolecules, 37(10), 3635-3641 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service