Skip to Content
Merck
All Photos(1)

Key Documents

434248

Sigma-Aldrich

L-Abrine

99%

Synonym(s):

Nα-Methyl-L-tryptophan

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$395.00

$395.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$395.00

About This Item

Empirical Formula (Hill Notation):
C12H14N2O2
CAS Number:
Molecular Weight:
218.25
Beilstein:
86638
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.22

$395.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

optical activity

[α]20/D +65°, c = 1 in 0.5 M NaOH

reaction suitability

reaction type: solution phase peptide synthesis

mp

>300 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

CN[C@@H](Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C12H14N2O2/c1-13-11(12(15)16)6-8-7-14-10-5-3-2-4-9(8)10/h2-5,7,11,13-14H,6H2,1H3,(H,15,16)/t11-/m0/s1

InChI key

CZCIKBSVHDNIDH-NSHDSACASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

An indoleamino acid that shows radical scavenging and antioxidant properties in vitro.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maximilian Huber et al.
The Science of the total environment, 541, 895-919 (2015-10-09)
A dataset of 294 monitored sites from six continents (Africa, Asia, Australia, Europe, North and South America) was compiled and evaluated to characterize the occurrence and fate of heavy metals in eight traffic area categories (parking lots, bridges, and three
Emilie Gauthy et al.
PloS one, 8(9), e76186-e76186 (2013-10-08)
GARP is a transmembrane protein present on stimulated human regulatory T lymphocytes (Tregs), but not on other T lymphocytes (Th cells). It presents the latent form of TGF-β1 on the Treg surface. We report here that GARP favors the cleavage
Huixi Zou et al.
Journal of natural products, 72(1), 44-52 (2008-12-31)
This study reports that a series of tryptophan derivatives with modifications on the side chain or at the indole ring were accepted by two cyclic dipeptide prenyltransferases, CdpNPT and FtmPT1, and converted to prenylated derivatives. The structures of the enzymatic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service