Skip to Content
Merck
All Photos(1)

Key Documents

431338

Sigma-Aldrich

Ammonium cerium(IV) nitrate

99.99%

Synonym(s):

Ceric ammonium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NH4)2(NO3)6
CAS Number:
Molecular Weight:
548.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.99%

reaction suitability

reagent type: oxidant

impurities

≤0.05% insol. dil. H2SO4
<100 ppm total metallic impurities

anion traces

chloride (Cl-): ≤0.01%
phosphate (PO43-): ≤0.02%

cation traces

Fe: ≤0.005%

SMILES string

N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ce.2HNO3.4NO3.2H3N/c;6*2-1(3)4;;/h;2*(H,2,3,4);;;;;2*1H3/q+4;;;4*-1;;

InChI key

WIBGOERAEYJBOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Offers a novel route to tartronic acid derivatives via facile oxygenation of malonates.

Features and Benefits

Meets ACS reagent specifications.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nair, V. et al.
Tetrahedron Letters, 39, 2801-2801 (1998)
Yue Qi Ye et al.
Bioscience, biotechnology, and biochemistry, 74(11), 2342-2344 (2010-11-13)
The structure of a long-known natural pigment, atromentin, was established by a total synthesis based on double Suzuki-Miyaura coupling and by a single-crystal X-ray analysis of the synthetic sample thereby obtained. A similar strategy including ceric ammonium nitrate (CAN) oxidation
D Subhas Bose et al.
Journal of combinatorial chemistry, 12(1), 100-110 (2009-12-17)
An efficient and practical method has been manifested for the diversity-oriented synthesis of quinolines via Friedländer annulation reaction for the generation of a wide range of structurally interesting and pharmacologically significant compounds by using ceric ammonium nitrate as a catalyst
S Sudha et al.
Ultrasonics sonochemistry, 19(5), 994-998 (2012-03-09)
Tetrahydrobenzo[c]xanthenes-11-ones was synthesized by a three component reaction of α-naphthol, aromatic aldehyde and dimedone. Ceric ammonium nitrate acts as a suitable eco-friendly catalyst for this method. Shorter reaction duration, mild reaction condition and low cost make this protocol more effective.
Weibin Song et al.
Bioorganic & medicinal chemistry letters, 22(1), 387-390 (2011-11-26)
A new, practical and efficient method for the synthesis of anhydrovinblastine AVBL (1f) by oxidative coupling of vindoline and catharanthine in the presence of ceric ammonium nitrate (CAN) was developed. Under the optimized reaction conditions, we synthesized a new series

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service