Skip to Content
Merck
All Photos(1)

Key Documents

429279

Sigma-Aldrich

Nylon 6/12

pellets

Synonym(s):

Hexamethylenediamine-1,12-dodecanedioic acid polymer, Poly(hexamethylene dodecanediamide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H22O4 · C6H16N2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002

abrasion resistance

6 mg (Taber, CS-17 wheel, 1kg, 50% RH, 1,000 cycles)

form

pellets

contains

lubricant

hardness

114 (Rockwell R, DAM, ASTM D 785)

mp

218 °C (lit.)

solubility

strong proton donor solvents, acetic acid and phenols: soluble

density

1.3 g/mL at 25 °C (lit.)

SMILES string

NCCCCCCN.OC(=O)CCCCCCCCCCC(O)=O

InChI

1S/C12H22O4.C6H16N2/c13-11(14)9-7-5-3-1-2-4-6-8-10-12(15)16;7-5-3-1-2-4-6-8/h1-10H2,(H,13,14)(H,15,16);1-8H2

InChI key

ZMUCVNSKULGPQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

General purpose thermoplastic polyamide.
Fibers for textiles, brushes and sutures cable sheathing and tubing and bearings, cams, gears and casings for tools and appliances.

Features and Benefits

Stiff and strong over a wide range of temperatures, excellent toughness and chemical and abrasion resistance. Attacked by strong acids and strong bases at elevated temperatures. Oxidized by chlorine and hydrogen peroxide.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service