Skip to Content
Merck
All Photos(1)

Documents

429155

Sigma-Aldrich

(Acetylacetonato)(1,5-cyclooctadiene)iridium(I)

Synonym(s):

Ir(acac)(COD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19IrO2
CAS Number:
Molecular Weight:
399.51
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder or crystals

reaction suitability

core: iridium
reagent type: catalyst

mp

150-155 °C (dec.)

SMILES string

CC(=O)\C=C(\C)O[Ir].C1CC=CCCC=C1

InChI

1S/C8H12.C5H8O2.Ir/c1-2-4-6-8-7-5-3-1;1-4(6)3-5(2)7;/h1-2,7-8H,3-6H2;3,6H,1-2H3;/q;;+1/p-1/b2-1-,8-7-;4-3-;

InChI key

JHNBKYKCHXHKET-DWVXZKBMSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Sava et al.
Chemico-biological interactions, 45(1), 1-6 (1983-07-01)
The effects of square planar rhodium, [RhacacCOD]o and iridium, [IracacCOD]o complexes and of octahedral ruthenium, [cis-RuCl2 (DMSO)4]o complex have been examined in comparison with cis-dichlorodiammino platinum(II) (cis-PDD). The toxicity in BDF1 mice varies widely and decreasing LD50-values, ranging from 0.94

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service