Skip to Content
Merck
All Photos(1)

Key Documents

427039

Sigma-Aldrich

Ethanol-13C2

99 atom % 13C

Synonym(s):

Ethyl alcohol-13C2

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$792.00
1 G
$2,390.00

$792.00


Check Cart for Availability

Request more information

Select a Size

Change View
250 MG
$792.00
1 G
$2,390.00

About This Item

Linear Formula:
13CH313CH2OH
CAS Number:
Molecular Weight:
48.05
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

$792.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.362 (lit.)

bp

78 °C (lit.)

mp

-114 °C (lit.)

density

0.839 g/mL at 25 °C

mass shift

M+2

storage temp.

room temp

SMILES string

[13CH3][13CH2]O

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3/i1+1,2+1

InChI key

LFQSCWFLJHTTHZ-ZDOIIHCHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mukund P Srinivasan et al.
Biochemical pharmacology, 180, 114174-114174 (2020-07-28)
Primary toxicity targets of alcohol and its metabolites in the pancreas are cellular energetics and endoplasmic reticulum (ER). Therefore, the role of AMP-Activated Protein Kinase (AMPKα) in amelioration of ethanol (EtOH)-induced pancreatic acinar cell injury including ER/oxidative stress, inflammatory responses

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service